Anti-CD58 Mouse Monoclonal Antibody (Biotin) [clone: MEM-63]
Catalog # ABNOMAB5005
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD58 Molecule
- Antigen symbol:CD58
- Clonality:Monoclonal
- Clone:MEM-63
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Pig,Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:965
- Antigen synonyms:LFA-3|LFA3|ag3
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CD58 from NALM-6 human pre-B cell line.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native CD58.
Recommended Dilutions: Flow Cytometry: 1:200
Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: MEM-63
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig