Order Entry
Poland
ContactUsLinkComponent
Anti-TNFRSF10A Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: DR-4-02]
Anti-TNFRSF10A Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: DR-4-02]
Catalog # ABNOMAB5076
Supplier:  Abnova
Anti-TNFRSF10A Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: DR-4-02]
Catalog # ABNOMAB5076
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tumor Necrosis Factor Receptor Superfamily, Member 10A
  • Antigen symbol:
    TNFRSF10A
  • Clonality:
    Monoclonal
  • Clone:
    DR-4-02
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    8797
  • Antigen synonyms:
    TRAILR1|DR4|APO2|CD261|TRAILR-1
  • Storage buffer:
    PBS (0,2% BSA, 15 mM sodium azide)
  • Storage temperature:
    Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant Fc fusion protein corresponding to extracellular region of TNFRSF10A.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant TNFRSF10A.

Recommended Dilutions: Flow Cytometry: 5 ?g/ml

Type: Primary
Antigen: TNFRSF10A
Clonality: Monoclonal
Clone: DR-4-02
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human