Anti-Benzo[a]pyrene Monoclonal Antibody [clone: BAP-13]
ABNOMAB3600
: Abnova
- Antibody type:Primary
- Antigen name:Benzo[a]pyrene
- Antigen symbol:Benzo[a]pyrene
- Clonality:Monoclonal
- Clone:BAP-13
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG1
- Reactivity:Bovine,Rat
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Benzo[a]pyrene conjugated with BSA.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against Benzo[a]pyrene.
Type: Primary
Antigen: Benzo[a]pyrene
Clonality: Monoclonal
Clone: BAP-13
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Bovine, Rat