Order Entry
Poland
ContactUsLinkComponent
 
Anti-HLA-DQ1/DQ3 Mouse Monoclonal Antibody [clone: HL-37]
Catalog # ABNOMAB3615
Supplier:  Abnova
Anti-HLA-DQ1/DQ3 Mouse Monoclonal Antibody [clone: HL-37]
Catalog # ABNOMAB3615
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HLA-DQ1/DQ3
  • Clonality:
    Monoclonal
  • Clone:
    HL-37
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG3
  • Reactivity:
    Pig,
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Antigen synonyms:
    DQ3
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native Burkitt's lymphoma cell line Raji.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native HLA-DQ1/DQ3.

Type: Primary
Antigen: HLA-DQ1/DQ3
Clonality: Monoclonal
Clone: HL-37
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Pig