Anti-S100 A9 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12045
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:S100 calcium binding protein A9
- Antigen symbol:S100A9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Antiserum
- Gene ID:6280
- Antigen synonyms:S100A9|S100 Calcium binding protein A9|CAGB|MRP14|CFAG|S100 calcium-binding protein A9|NIF|MAC387|MIF|L1AG|calgranulin B|60B8AG|CGLB|P14|S100 calcium-binding protein A9 (calgranulin B)|LIAG|S100 Calcium binding protein A9 (calgranulin B)
- Storage buffer:0,1% sodium azide
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human S100A9.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against full length recombinant S100A9.
Recommended Dilutions: Immunocytochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200; Western Blot: 1:1000
Type: Primary
Antigen: S100 A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat