Anti-NDUFAF3 Rabbit Polyclonal Antibody
numer katalogowy ABNOPAB22870
Dostawca: Abnova
Dane techniczne
- Antibody type:Primary
- Antigen name:NADH dehydrogenase (ubiquinone) complex I, assembly factor 3
- Antigen symbol:NDUFAF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:25915
- Antigen synonyms:wu:fc05g04|zgc:153176
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:LLEPRIEIVVVGTGDRTERLQSQVLQAMRQRGIAVEVQDTPNACATFNFLCHEGRVTGAALIPPPGGTSLTSLGQAAQ
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human NDUFAF3.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Dane techniczne
Informacje o tym produkcie
Rabbit polyclonal antibody raised against recombinant NDUFAF3.
Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500
Type: Primary
Antigen: NDUFAF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human