Anti-VIM Mouse Monoclonal Antibody [clone: VI-01]
Catalog # ABNOMAB3880
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Vimentin
- Antigen symbol:VIM
- Clonality:Monoclonal
- Clone:VI-01
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Pig,Mammals
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:407747
- Antigen synonyms:CTRCT30|HEL113
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Native purified VIM from porcine brain cold stable proteins after depolymerization of microtubules.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native VIM.
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: VI-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mammals, Pig