Anti-CSF2 Mouse Monoclonal Antibody [clone: 29 (formerly 19)]
Catalog # ABNOMAB6267
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Colony Stimulating Factor 2
- Antigen symbol:CSF2
- Clonality:Monoclonal
- Clone:29 (formerly 19)
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:1437
- Antigen synonyms:GMCSF|TUBB2|Csfgm|Gm-CSf|TUBB|MGI-IGM|CDCBM5
- Storage buffer:10 mM PBS, pH 7,2
- Storage temperature:Store at –20 °C. Aliquot after reconstitution to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human CSF2.
- Size:500 μg
- Pk:500 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant CSF2.
Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: 29 (formerly 19)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human