Anti-IgE Mouse Monoclonal Antibody [clone: 4F4]
Catalog # ABNOMAB8447
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Immunoglobulin E
- Antigen symbol:IgE
- Clonality:Monoclonal
- Clone:4F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at 4 °C.
- Immunogen:Native purified IgE from human myeloma patient serum.
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Mouse monoclonal antibody raised against native IgE.
Type: Primary
Antigen: IgE
Clonality: Monoclonal
Clone: 4F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human