Order Entry
Poland
ContactUsLinkComponent
Anti-IgG1 Mouse Monoclonal Antibody [clone: MOPC-21]
Anti-IgG1 Mouse Monoclonal Antibody [clone: MOPC-21]
Catalog # ABNOMAB8514
Supplier:  Abnova
Anti-IgG1 Mouse Monoclonal Antibody [clone: MOPC-21]
Catalog # ABNOMAB8514
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Immunoglobulin G1
  • Antigen symbol:
    IgG1
  • Clonality:
    Monoclonal
  • Clone:
    MOPC-21
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against IgG1.

Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse