Anti-GDF15 Rabbit Polyclonal Antibody
ABNOPAB16899
: Abnova
- Antibody type:Primary
- Antigen name:Growth differentiation factor 15
- Antigen symbol:GDF15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:9518
- Antigen synonyms:MIC-1|GDF-15|NAG-1|PTGFB|PDF|PLAB|MIC1
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human GDF15.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against full length recombinant GDF15.
Recommended Dilutions: ELISA: 1:500; Western Blot: 1:800-1:1500
Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse