Order Entry
Poland
ContactUsLinkComponent
Anti-Leishmania A2 Mouse Monoclonal Antibody [clone: C9]
Anti-Leishmania A2 Mouse Monoclonal Antibody [clone: C9]
Catalog # ABNOMAB5675
Supplier:  Abnova
Anti-Leishmania A2 Mouse Monoclonal Antibody [clone: C9]
Catalog # ABNOMAB5675
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Leishmania A2
  • Clonality:
    Monoclonal
  • Clone:
    C9
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Protozoa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Store at 4 °C or –20 °C on dry atmosphere. After reconstitution with water and glycerol (1:1), store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native from A2 protein of Leishmania donovani and Leishmania infantum.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against A2.

Recommended Dilutions: Western Blot: 1:100

Type: Primary
Antigen: Leishmania A2
Clonality: Monoclonal
Clone: C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Protozoa