Anti-KCNG1 Rabbit Polyclonal Antibody
ABNOPAB23612
: Abnova
- Antibody type:Primary
- Antigen name:Potassium Voltage-gated Channel Subfamily G Member 1
- Antigen symbol:KCNG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:3755
- Antigen synonyms:KCNG1_HUMAN.|Potassium voltage gated channel subfamily G|K13|KCNG|kH2|Voltage gated potassium channel subunit Kv6.1|Potassium voltage gated channel subfamily G member 1|KV6.1
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:ELKQEQERVMFRRAQFLIKTKSQLSVSQDSDILFGSASSDTRDNN
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human KCNG1.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against recombinant KCNG1.
Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Immunofluorescence: 1-4 ?g/ml
Type: Primary
Antigen: KCNG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human