Anti-SHANK2 Mouse Monoclonal Antibody [clone: S23B-49]
Catalog # ABNOMAB7890
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:SH3 and multiple ankyrin repeat domains 2
- Antigen symbol:SHANK2
- Clonality:Monoclonal
- Clone:S23B-49
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:171093
- Antigen synonyms:AUTS17|CORTBP1|ProSAP1|SHANK|CTTNBP1|SPANK-3
- Amino acid number:84 to 309
- Storage buffer:PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 84-309 of rat Shank2.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant Shank2.
Recommended Dilutions: Western Blot: 1-10 ?g/ml; Immunohistochemistry: 0.1-1.0 ?g/ml; Immunocytochemistry: 0.1-1.0 ?g/ml; Immunofluorescence: 1-10 ?g/ml
Type: Primary
Antigen: SHANK2
Clonality: Monoclonal
Clone: S23B-49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat