Order Entry
Poland
ContactUsLinkComponent
 
Anti-H2-D4 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 34-5-8S]
  ABNOMAB5820
 :  Abnova
Anti-H2-D4 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 34-5-8S]
  ABNOMAB5820
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen symbol:
    H2-D4
  • Clonality:
    Monoclonal
  • Clone:
    34-5-8S
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS (1 mg/mL sodium azide)
  • Storage temperature:
    Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    C57BL/6 x DBA/2)F1 hybrid mouse splenocytes.
  • Size:
    500 μg
  • Pk:
    500 µg

 

 

Mouse monoclonal antibody raised against H2-D4.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: H2-D4
Clonality: Monoclonal
Clone: 34-5-8S
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse