Anti-H2-D2 Mouse Monoclonal Antibody (Biotin) [clone: 27-11-13S]
ABNOMAB5824
: Abnova
- Antibody type:Primary
- Antigen symbol:H2-D2
- Clonality:Monoclonal
- Clone:27-11-13S
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (<
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:C3H alphaBDF1 splenocytes.
- Size:500 μg
- Pk:500 µg
Mouse monoclonal antibody raised against H2-D2.
Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells
Type: Primary
Antigen: H2-D2
Clonality: Monoclonal
Clone: 27-11-13S
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse