Order Entry
Poland
ContactUsLinkComponent
Anti-H2-D2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 27-11-13S]
Anti-H2-D2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 27-11-13S]
Catalog # ABNOMAB5825
Supplier:  Abnova
Anti-H2-D2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 27-11-13S]
Catalog # ABNOMAB5825
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    H2-D2
  • Clonality:
    Monoclonal
  • Clone:
    27-11-13S
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS (0,1 mg/mL sodium azide)
  • Storage temperature:
    Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    C3H alphaBDF1 splenocytes.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against H2-D2.

Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells

Type: Primary
Antigen: H2-D2
Clonality: Monoclonal
Clone: 27-11-13S
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse