Order Entry
Poland
ContactUsLinkComponent
Anti-Hepatitis C virus core Protein Mouse Monoclonal Antibody [clone: H6-29]
Anti-Hepatitis C virus core Protein Mouse Monoclonal Antibody [clone: H6-29]
Catalog # ABNOMAB7949
Supplier:  Abnova
Anti-Hepatitis C virus core Protein Mouse Monoclonal Antibody [clone: H6-29]
Catalog # ABNOMAB7949
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Hepatitis C virus core Protein
  • Clonality:
    Monoclonal
  • Clone:
    H6-29
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Virus
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Amino acid number:
    13 to 124
  • Storage buffer:
    PBS (50% glycerol)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids 13-124 of HCV core protein.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against recombinant hepatitis C virus core protein.

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: H6-29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Virus