Anti-ATOX1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB15530
Supplier: Abnova
Specifications
- Typ przeciwciała:Primary
- Nazwa antygenu:Antioxidant 1 Copper Chaperone
- Symbol antygenu:ATOX1
- Klonalność:Poliklonalne
- Koniugacja:Unconjugated
- Gospodarz:Rabbit
- Reaktywność:Human
- Western blot:Yes
- Adsorpcja krzyżowa:No
- Format:Liquid
- Identyfikator genu:475
- Synonimy antygenów:HAH1|ATX1
- Bufor do przechowywania:PBS (0,1% sodium azide)
- Temperatura przechowywania:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Full length human ATOX1.
- Rozmiar:100 μl
- Ilość:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against ATOX1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: ATOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human