Order Entry
Poland
ContactUsLinkComponent
Anti-Rabies virus Mouse Monoclonal Antibody
Anti-Rabies virus Mouse Monoclonal Antibody
Catalog # ABNOMAB8251
Supplier:  Abnova
Anti-Rabies virus Mouse Monoclonal Antibody
Catalog # ABNOMAB8251
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Rabies virus
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS (0,1% sodium azide)
  • Storage temperature:
    Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Rabies virus.
  • Size:
    200 μg
  • Pk:
    200 µg

Specifications

About this item

Mouse monoclonal antibody raised against rabies virus.

Recommended Dilutions: Immunofluorescence: 1:1000

Type: Primary
Antigen: Rabies virus
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: