Anti-Clostridium difficile Cell Surface Protein Rabbit Polyclonal Antibody
ABNOPAB16014
: Abnova
- Antibody type:Primary
- Antigen name:Clostridium difficile Cell Surface Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Clostridium spp.
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,2 (50% glycerol, 0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus of Clostridium difficile Cell Surface Protein.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of Clostridium difficile Cell Surface Protein.
Recommended Dilutions: ELISA: 1:2000-1:5000; Western Blot: 1 ?g/ml
Type: Primary
Antigen: Clostridium difficile Cell Surface Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Clostridium Species