Anti-CHRNB2 Goat Polyclonal Antibody
Catalog # ABNOPAB18684
Dostawca: Abnova
Dane techniczne
- Antibody type:Primary
- Antigen name:Cholinergic receptor, nicotinic, beta 2 (neuronal)
- Antigen symbol:CHRNB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Cross adsorption:No
- Format:Liquid
- Gene ID:1141
- Antigen synonyms:nicotinic|beta 2 (neuronal) |cholinergic receptor|EFNL3|nAChRB2|beta polypeptide 2 (neuronal)|neuronal nicotinic acetylcholine receptor beta 2|CHRNB2
- Storage buffer:Tris saline, pH 7,3 (0,02% sodium azide, 0,5% BSA)
- Sequence:QPRHHCARQRLR
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at internal region of human CHRNB2.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Dane techniczne
Informacje o tym produkcie
Goat polyclonal antibody raised against synthetic peptide of CHRNB2.
Recommended Dilutions: ELISA: 1:64000; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: CHRNB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human