Order Entry
Poland
ContactUsLinkComponent
IgG Isotype Control (APC (Allophycocyanin)/Cy5.5®)
IgG Isotype Control (APC (Allophycocyanin)/Cy5.5®)
Catalog # ABNOPAB19467
Supplier:  Abnova
IgG Isotype Control (APC (Allophycocyanin)/Cy5.5®)
Catalog # ABNOPAB19467
Supplier:  Abnova

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Conjugation:
    APC (Allophycocyanin)/Cy5.5®
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Isotype:
    IgG
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS (with sodium azide)
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse IgG raised from normal mouse serum.

Recommended Dilutions: Flow Cytometry: <1 ?g/106 cells; ELISA: 1:500-1:5000

Type: Control
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: APC (Allophycocyanin)/Cy5.5®
Epitope:
Host:
Isotype: IgG
Reactivity: