Anti-Dengue virus E-D2 Mouse Monoclonal Antibody [clone: 7u85h]
Catalog # ABNOMAB8904
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Dengue virus E-D2
- Clonality:Monoclonal
- Clone:7u85h
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Virus
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Amino acid number:52 to 280
- Storage buffer:citrate-Tris-HCl, pH7.0 (0,02% Procl300)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 52-280 of Dengue virus E-D2.
- Purification:Affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant Dengue virus E-D2.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: Dengue virus E-D2
Clonality: Monoclonal
Clone: 7u85h
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Virus