IgG Isotype Control (FITC (Fluorescein Isothiocyanate))
ABNOPAB19457
: Abnova
- Antibody type:Control
- Clonality:Monoclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Isotype:IgG
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (with sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Size:500 μg
- Pk:500 µg
Mouse IgG raised from normal mouse serum.
Recommended Dilutions: Flow Cytometry: <1 ?g/106 cells; ELISA: 1:500-1:5000
Type: Control
Antigen: IgG
Clonality: FITC (Fluorescein Isothiocyanate)
Clone: Monoclonal
Conjugation:
Epitope:
Host: IgG
Isotype:
Reactivity: