IgG Isotype Control (RPE (R-Phycoerythrin)/Cy7®)
ABNOPAB19466
: Abnova
- Antibody type:Control
- Clonality:Monoclonal
- Conjugation:RPE (R-Phycoerythrin)/Cy7®
- ELISA:Yes
- Flow cytometry:Yes
- Isotype:IgG
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (with sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Size:100 μg
- Pk:100 µg
Mouse IgG raised from normal mouse serum.
Recommended Dilutions: Flow Cytometry: <1 ?g/106 cells; ELISA: 1:500-1:5000
Type: Control
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)/Cy7®
Epitope:
Host:
Isotype: IgG
Reactivity: