Order Entry
Poland
ContactUsLinkComponent
Anti-COX4I1 Goat Polyclonal Antibody
Anti-COX4I1 Goat Polyclonal Antibody
Catalog # ABNOPAB7322
Supplier:  Abnova
Anti-COX4I1 Goat Polyclonal Antibody
Catalog # ABNOPAB7322
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytochrome C Oxidase Subunit Iv Isoform 1
  • Antigen symbol:
    COX4I1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    1327|84701
  • Antigen synonyms:
    COXIV|IV-1|AL024441|COX|COX IV-1|Cox4a|Cox4
  • Storage buffer:
    Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
  • Sequence:
    C-QGLASKWDYEKNE
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to human COX4I1/COX4I2.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Goat polyclonal antibody raised against synthetic peptide of COX4I1/COX4I2.

Recommended Dilutions: ELISA: 1:4000; Western Blot: 0.01-0.03 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml

Type: Primary
Antigen: COX4I1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human