Order Entry
Poland
ContactUsLinkComponent
 
Anti-Collagen type VI Rabbit Polyclonal Antibody
  ABNOPAB19146
 :  Abnova
Anti-Collagen type VI Rabbit Polyclonal Antibody
  ABNOPAB19146
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Collagen type VI
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Marmoset,
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS
  • Storage temperature:
    Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified collagen type VI from human placenta.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Rabbit polyclonal antibody raised against native collagen type VI.

Recommended Dilutions: Western Blot: 200-800 ng/ml; Immunohistochemistry: 250-800 ng/ml; Immunofluorescence: 300-900 ng/ml

Type: Primary
Antigen: Collagen type VI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Marmoset, Rat