Anti-STK4 Rabbit Polyclonal Antibody
Catalog # ABNOPAB18249
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Serine/threonine Kinase 4
- Antigen symbol:STK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:6789|6788
- Antigen synonyms:YSK3|TIIAC|MST1|KRS2
- Storage buffer:PBS, 150mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to residues surrounding T183 of human STK4/STK3.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of STK4/STK3.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:100; ELISA: 1:5000
Type: Primary
Antigen: STK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse