Anti-ADAM20 Rabbit Polyclonal Antibody
Catalog # ABNOPAB19740
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase Domain 20
- Antigen symbol:ADAM20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Serum
- Gene ID:8748
- Storage buffer:0,02% sodium azide
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 15 amino acids at C-terminus of human ADAM20.
- Purification:Protein A purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of ADAM20.
Recommended Dilutions: ELISA: 1:160000; Western Blot: 1:500-1000
Type: Primary
Antigen: ADAM20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human