Anti-ABCE1 Goat Polyclonal Antibody
ABNOPAB6847
: Abnova
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, Subfamily E, Member 1
- Antigen symbol:ABCE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:6059
- Antigen synonyms:RNase 4 inhibitor|Ribonuclease 4 Inhibitor|ABC 38|ABCE 1|OABP|ATP Binding Cassette Sub Family E Member 1|RLI|RNS4I|ABC38|RNase L Inhibitor
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-KLNSIKDVEQKK
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human ABCE1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of ABCE1.
Recommended Dilutions: ELISA: 1:8000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: ABCE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human