Anti-HAVCR2 Goat Polyclonal Antibody
ABNOPAB6962
: Abnova
- Antibody type:Primary
- Antigen name:Hepatitis A virus cellular receptor 2
- Antigen symbol:HAVCR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:84868
- Antigen synonyms:kidney injury molecule 3|KIM-3|TIMD3|FLJ14428|hepatitis A virus cellular receptor 2|T cell immunoglobulin mucin 3|TIM3|HAVCR2
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-KWYSHSKEKIQN
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human HAVCR2.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of HAVCR2.
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: HAVCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human