Anti-HA tag Polyclonal Antibody
ABNOPAB7703
: Abnova
- Antibody type:Primary
- Antigen name:HA tag
- Antigen symbol:HA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
- Sequence:YPYDVPDYA
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to HA tag.
- Purification:Immunoaffinity purification
- Pk:100 µl
Rabbit polyclonal antibody raised against synthetic peptides of HA (Hemagglutinin) tag.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: HA tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: