Anti-AGAP2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB9929
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ArfGAP with GTPase domain, ankyrin repeat and PH domain 2
- Antigen symbol:AGAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:116986
- Antigen synonyms:GGAP2|CENTG1|PIKE
- Amino acid number:1 to 836
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C on dry atmosphere. After reconstitution with 0.1 mL of deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 1-836 of human AGAP2.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against partial recombinant AGAP2.
- Recommended Dilutions: ELISA: 1:40000-1:200000; Western Blot: 1:5000-1:20000
Type: Primary
Antigen: AGAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat