Anti-HDAC5 Mouse Monoclonal Antibody [clone: 4G2]
ABNOH00010014-M01A
: Abnova
- Antibody type:Primary
- Antigen name:Histone deacetylase 5
- Antigen symbol:HDAC5
- Clonality:Monoclonal
- Clone:4G2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites fluid
- Gene ID:10014
- Antigen synonyms:HDAC5|Antigen NY-CO-9|KIAA0600|HD5
- Amino acid number:330 to 429
- Sequence:AENGFTGSVPNIPTEMLPQHRALPLDSSPNQFSLYTSPSLPNISLGLQATVTVTNSHLTASPKLSTQQEAERQALQSLRQGGTLTGKFMSTSSIPGCLLG
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:HDAC5 (AAH51824, 330 a.a. ~ 429 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:200 μl
- Pk:200 µl
Mouse monoclonal antibody raised against a partial recombinant HDAC5.
Type: Primary
Antigen: HDAC5
Clonality: Monoclonal
Clone: 4G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human