Anti-CD14 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 61D3]
Supplier: Tonbo Biosciences
Certificates
About this item
The 61D3 antibody is specific for human CD14, a 53-55 kDa GPI-anchored glycoprotein. CD14 is highly expressed on monocytes and to a lesser extent on interfollicular macrophages, and some dendritic cells. Together with LPS-Binding Protein (LBP), CD14 binds to and mediates the innate immune response to bacterial lipopolysaccharide (LPS).
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Specifications
- Catalog No:
- 35-0149-T100
- 35-0149-T025
- Antigen symbol:
- CD14
- CD14
- Antigen name:
- CD14 Molecule
- CD14 Molecule
- Antigen synonyms:
- monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
- monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 61D3
- 61D3
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 929
- 929
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- +2 °C to +8 °C, protected from light
- +2 °C to +8 °C, protected from light
Specifications
Frequently Bought Together