Rat IgG2b Isotype Control [clone: LTF-2] (redFluor® 710)
Catalog # 80-4031-U100
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Control
- Clone:LTF-2
- Conjugation:redFluor® 710
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Neutralisation:Yes
- Storage temperature:Store Undiluted at 2 to 8 °C
- Size:100 μg
- Pk:100 µG
Specifications
About this item
The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.
Type: Control
Antigen: LTF-2
Clonality:
Clone: LTF-2
Conjugation: redFluor™ 710
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: N/A