Order Entry
Poland
ContactUsLinkComponent
Anti-BBS7 Rabbit Polyclonal Antibody
Anti-BBS7 Rabbit Polyclonal Antibody
numer katalogowy ABGEAP17417A
Supplier:  Abgent
Anti-BBS7 Rabbit Polyclonal Antibody
numer katalogowy ABGEAP17417A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Bardet-Biedl syndrome 7
  • Antigen symbol:
    BBS7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Gene ID:
    55212
  • Antigen synonyms:
    BBS2L1
  • Amino acid number:
    73 to 99
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    80.353 kDa
  • Sequence:
    SEIRGFTKRGKQC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.7
  • Shipping temperature:
    4 °C
  • Immunogen:
    This BBS7 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 73-99 amino acids from the N-terminal region of human BBS7.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: BBS7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human