Anti-Cytokeratin Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: C-11]
Catalog # CAYM10349-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin
- Clonality:Monoclonal
- Clone:C-11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Storage buffer:PBS, pH 7,2, containing 50% glycerol, 0,5 mg/ml BSA, and 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: Cytokeratin
Clonality: Monoclonal
Clone: C-11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human