Order Entry
Poland
ContactUsLinkComponent
Anti-Theophylline Sheep Polyclonal Antibody
Anti-Theophylline Sheep Polyclonal Antibody
Catalog # OXBTOBT1432
Anti-Theophylline Sheep Polyclonal Antibody
Catalog # OXBTOBT1432

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Theophylline
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • Isotype:
    IgG
  • Pk:
    1 mL

Specifications

About this item

Type: Primary
Antigen: Theophylline
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: chemical