Order Entry
Poland
ContactUsLinkComponent
Anti-DNP Rat Polyclonal Antibody
Anti-DNP Rat Polyclonal Antibody
Catalog # OXBTOBT1139
Anti-DNP Rat Polyclonal Antibody
Catalog # OXBTOBT1139

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    DNP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rat
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: DNP
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: