Anti-STAT5A Mouse Monoclonal Antibody (Unconjugated) [clone: 5F6.F1]
Catalog # ROCK200-301-A45S
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Signal transducer and activator of transcription 5A
- Antigen symbol:STAT5A
- Clonality:Monoclonal
- Clone:5F6.F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:20850
- Antigen synonyms:AA959963|STAT5
- Modification:phospho-Tyr694
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-STAT5 phospho Y694 monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding Y694 of mouse STAT5a Protein.
- Purification:Protein A purified
- Size:25 µl
- Pk:25 µl
Specifications
About this item
This monoclonal antibody is suitable for ELISA, immunochemistry and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 91 kDa in size.
Recommended Dilutions: ELISA: 1:20000; Immunofluorescence: 1:50 - 1:1000; Western Blot: 1:500 - 1:2000; Immunochemistry: 20-40 µg/ml;
Type: Primary
Antigen: STAT5A
Clonality: Monoclonal
Clone: 5F6.F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Mouse