Order Entry
Poland
ContactUsLinkComponent
Anti-KLH Mouse Monoclonal Antibody (Unconjugated) [clone: 12B4.G3.A8]
Anti-KLH Mouse Monoclonal Antibody (Unconjugated) [clone: 12B4.G3.A8]
Catalog # ROCK200-301-466
undefined
Anti-KLH Mouse Monoclonal Antibody (Unconjugated) [clone: 12B4.G3.A8]
Catalog # ROCK200-301-466

Specifications

  • Antibody type:
    Tag
  • Antigen symbol:
    KLH
  • Clonality:
    Monoclonal
  • Clone:
    12B4.G3.A8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Sequence:
    SLAFEEGSpQSTTISS
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    Anti-KLH (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with KLH (Keyhole Limpet Hemocyanin) followed by hybridoma development.
  • Purification:
    Protein A purified
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Anti-KLH can be used as a control in various assays or in ELISA assays to detect KLH or KLH conjugates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000

Type: Tag
Antigen: KLH
Clonality: Monoclonal
Clone: 12B4.G3.A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Klh,