Order Entry
Poland
ContactUsLinkComponent
 
Anti-NXN Rabbit Polyclonal Antibody
  USBI151996-200
Anti-NXN Rabbit Polyclonal Antibody
  USBI151996-200

 

  • Antibody type:
    Primary
  • Antigen name:
    Nucleoredoxin
  • Antigen symbol:
    NXN
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Purification:
    Affinity Purified
  • Pk:
    200 µl

 

 

Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: NXN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human