Anti-LAT Mouse Monoclonal Antibody [clone: 11B.12]
USBIL1390-01B-100
- Antibody type:Primary
- Antigen symbol:LAT
- Clonality:Monoclonal
- Clone:11B.12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Amino acid number:31 to 233
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,5 mg/ml
- Immunogen:GST fusion protein corresponding to aa31-233 of human LAT, cytoplasmic domain.
- Purification:purified by Protein G chromatography
- Size:100 μg
- Pk:100 µG
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: LAT
Clonality: Monoclonal
Clone: 11B.12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse,