Anti-Relaxin 1,2 Mouse Monoclonal Antibody [clone: 8H11kurd]
Catalog # USBIR1424-96C-10
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Relaxin 1,2
- Clonality:Monoclonal
- Clone:8H11kurd
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Human Relaxin 1/2
- Purification:purified by Protein G chromatography
- Size:10 μg
- Pk:10 µG
Specifications
About this item
Type: Primary
Antigen: Relaxin 1,2
Clonality: Monoclonal
Clone: 8H11kurd
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,