Order Entry
Poland
ContactUsLinkComponent
Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 21-46]
Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 21-46]
Catalog # USBIB2702-33D-10
Supplier:  US Biological
Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 21-46]
Catalog # USBIB2702-33D-10
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Brain Natriuretic Peptide
  • Antigen symbol:
    BNP
  • Clonality:
    Monoclonal
  • Clone:
    21-46
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    lyophilized powder
  • Amino acid number:
    1 to 32
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml
  • Immunogen:
    Synthetic human BNP (aa 1-32) KLH conjugated (SPKMVQGSGCFGRKMDRISSSSGLGCKVLRRH)
  • Purification:
    purified by Protein G chromatography
  • Size:
    10 μg
  • Pk:
    10 µG

Specifications

About this item

Type: Primary
Antigen: Brain Natriuretic Peptide
Clonality: Monoclonal
Clone: 21-46

Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,