Anti-GID2 Rabbit Polyclonal Antibody
AGRIAS132638
: Agrisera
- Antibody type:Primary
- Antigen symbol:GID2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Spinacia oleracea,C. reinhardtii,Brassica rapa var. komatsuna,Zea mays,A. thaliana,N. tabacum,Brassica oleracea var. italica,Citrus limon
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:Q9STX3
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana GID2 sequence,
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: GID2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Brassica oleracea var. italica, Brassica rapa var. komatsuna, C. reinhardtii, Citrus limon, N. tabacum, Spinacia oleracea, Zea mays