Anti-TPS1 Rabbit Polyclonal Antibody
Catalog # AGRIAS122635
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:TPS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:Q9SYM4
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana TPS1,
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: TPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana