Anti-V-PPase Rabbit Polyclonal Antibody
AGRIAS121849
: Agrisera
- Antibody type:Primary
- Antigen name:V-PPase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Picea abies,A. thaliana
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:P31414
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana V-PPase,UniProt P31414, TAIR AT1G15690
- Size:100 µl
- Pk:50 µl
Type: Primary
Antigen: V-PPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Picea abies